Learn More
Bioassay Systems QuantiChrom™ Urease Assay Kit. For quantitative determination of urease activity and evaluation/screen for urease inhibitors.
SDP
Supplier:  Bioassay Systems DURE100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For quantitative determination of urease activity and evaluation/screen for urease inhibitors. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.003 U/L urease activity can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD670nm; Samples: Biological, environment etc; Size: 100 tests.
Catalog No. 50-195-6295
missing translation for 'QuantityLeft'