Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10058-F4, Tocris Bioscience™

Catalog No. p-4815630 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits c-Myc-Max dimerization

10058-F4 is a cell permeable c-Myc-Max dimerization inhibitor. Inhibits proliferation, induces apoptosis and arrests cells in G0/G1 in rat1a-c-Myc cells. Also reduces tumor growth in vivo. In lymphoma cells, 10058-F4 decreases glucose uptake and expression of glycolysis-associated genes and reduces cell viability.

Chemical Identifiers

CAS 403811-55-2
Molecular Formula C12H11NOS2
Molecular Weight (g/mol) 249.346
InChI Key SVXDHPADAXBMFB-JXMROGBWSA-N
Synonym c-myc inhibitor, 5e-5-4-ethylbenzylidene-2-mercapto-1,3-thiazol-4 5h-one, 5-4-ethylphenyl methylene-2-thioxo-4-thiazolidinone, e-5-4-ethylbenzylidene-2-mercaptothiazol-4 5h-one, z,e-5-4-ethylbenzylidine-2-thioxothiazolidin-4-one, 5e-5-4-ethylphenyl methylidene-2-sulfanyl-1,3-thiazol-4-one, 5e-5-4-ethylphenyl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one, 10058-f hplc , solid
PubChem CID 1271002
IUPAC Name (5E)-5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CCC1=CC=C(C=C1)C=C2C(=O)NC(=S)S2

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 249.35
Percent Purity >99%
Physical Form Solid
Chemical Name or Material 10058-F4
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.