Learn More
Boston Bioproducts Inc 100ml Magnesium Sulfate, 1m
Supplier:  Boston Bioproducts Inc MT210100ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
1 M Magnesium sulfate solution. Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. NC9968111