Learn More
Ace Glass, Inc. 100mL volumetric bulb for use with ACE Automatic Dispensing Pipet.
Supplier: Ace Glass, Inc. 800445
100mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.