Learn More
SPEX CERTIPREP LLC 3P 100PPM PHOSPHOROUS KH2PO4 IC
Supplier:  SPEX CERTIPREP LLC 3P ICKP500
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Phosphorus from KH2PO4 in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9586382
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.