Learn More
Essendant Conqueror 103 Odor Counteractant Concentrate, Lemon, 1 gal Bottle, 4/Carton
Supplier: Essendant FRS1WBLE
This water-soluble formula cleans, deodorizes and refreshes surfaces. The formula contains powerful detergents and potent perfumes to counteract malodors and leave the area smelling fresh and clean. Add 1-2 oz. per gallon of water or cleaning solution. Use in fabric shampoo, mop water, any water-based solution. Can be used in trigger sprayers, carpet extractors and laundry.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.