Learn More
Ace Glass, Inc. 10MM POR B TUBES (GB)
Supplier: Ace Glass, Inc. 720504
10mm straight tubes with 70-100 micron fritted discs, for construction of special apparatus.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.