Learn More
Bioassay Systems QuantiQuik™ L-Lactic Acid Quick Test Strips.Direct determination of L-Lactate concentrations in food and beverage samples as well as biological samples.
SDP
Supplier:  Bioassay Systems QQLLAC10
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
L-LACTIC ACID, or L-lactate, is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. L-lactic acid is added to many foods and beverages to give it a tart flavor. Increased levels of L-lactic acid in milk, egg, and fruit juices can be an indication of spoilage. In the wine industry, increasing levels of L-lactic acid and the decreasing levels of L-malic acid are monitored (Malolactic fermentation). In this process, the overall acidity of the wine is reduced and can lead to the improvement of the flavor of the wine.BioAssay Systems QuantiQuik™ L-Lactic Acid Test Strips are based on L-lactate dehydrogenase catalyzed oxidation of L-lactate in which the formed NADH reduces a chromogenic reagent. The intensity of product color is directly proportional to L-lactate concentration in the sample.
Catalog No. 50-195-6344
missing translation for 'QuantityLeft'