Learn More
USA Scientific Inc .1-10ul flat gel tip, .37 mm
Supplier:  USA Scientific Inc 10124600
Encompass
10 uL flat sequencing gel tip, 0.37 mm OD orifice, 96 tips/rack, 2 racks/pack (192 tips)
Catalog No. 50-274-4997
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.