Learn More
AdipoGen 12-1-Pyrenyl dodecanoic acid
Supplier: AdipoGen CDXP0006M050
Chemical. CAS 69168-45-2. Formula C28H32O2. MW 400.55. Synthetic. Anionic membrane probe used for studies on the uptake of fluorescent fatty acids into cultured cells and oxygen sensing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.