Learn More
Medchemexpress LLC HY-112530 100mg Medchemexpress, 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine CAS:1069-79-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112530100MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-112530 100mg 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine CAS:1069-79-0 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid that can be used in the synthesis of liposomes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0819