Learn More
Chemglass Life Sciences 125mL Erlenmeyer Flask, Iodine, Stopper, COMPLETE
SDP
Supplier:  Chemglass Life Sciences CG155201
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
125mL Erlenmeyer Flask, Iodine, Stopper, COMPLETE. Flask has a funnel top with a standard taper outer grind to accept a 24/40 long neck stopper allowing the handle to extend above the liquid seal. Used in ASTM D29 and ASTM D555. Available complete with stopper or flask only.
Catalog No. CG155201