Learn More
Chemglass Life Sciences 125mL Erlenmeyer Flask, Iodine, Stopper, FLASK ONLY
SDP
Supplier:  Chemglass Life Sciences CG155202
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
125mL Erlenmeyer Flask, Iodine, Stopper, FLASK ONLY. Flask has a funnel top with a standard taper outer grind to accept a 24/40 long neck stopper allowing the handle to extend above the liquid seal. Used in ASTM D29 and ASTM D555. Available complete with stopper or flask only. THIS ITEM IS FLASK ONLY.
Catalog No. CG155202