Learn More
Ace Glass, Inc. 150mL jacketed filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 60mm IDx75mm depth, with CAPFE O-ring, 28/15 O-ring ball joint inlet/outlet connects, pressure rated at 35 psi.
Supplier:  Ace Glass, Inc. 638606
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
150mL jacketed filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 60mm IDx75mm depth, with CAPFE O-ring, 28/15 O-ring ball joint inlet/outlet connects, pressure rated at 35 psi.
Catalog No. 50-972-798