Learn More
Chemglass Life Sciences 15L Process Reactor, Complete, Air Motor, 67.0" OAH with Motor, 15.3" Clearance Below Valve
SDP
Supplier:  Chemglass Life Sciences CG1964615
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
15L Process Reactor, Complete, Air Motor, 67.0" OAH with Motor, 15.3" Clearance Below Valve. PLEASE NOTE: REACTOR STAND OVERALL HEIGHT WITH MOTOR AND CLEARANCE BELOW VALVE, ARE CALCULATED FOR ALL CHEMGLASS STIRRER BEARINGS.Features:• Support Frame with Oversized Rugged Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings• Improved Support Clamp Auto-Centers Reactor• Perfluoro O-Rings On 60mm and 200mm Flanges• Stainless Steel Inlet/Outlet Adapters Allow Connection To Cooling/Heating Circulator• Zero Dead Space Detachable Drain Valve Has Knob with Laser Etched Directional Arrows For Open and Close• Air Motor. Optional XP Motor Available• 5-Neck Lid with Large Powder Addition Funnel• Digital Temperature Monitor• Reactor Jacket Graduated On Both Sides• Removable Stainless Steel Pipe Used To Support Side Shelf and Manifold System• Support Bushings For Optional Safety Shield• Heavy Duty Lockable Casters
Catalog No. CG1964615
missing translation for 'QuantityLeft'