Learn More
Jackson Immuno Research Labs 18 nm Colloidal Gold AffiniPure Donkey Anti-Rabbit IgG (H+L) (EM Grade), 0.3mL
SDP
Supplier:  Jackson Immuno Research Labs 711215152
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective.
Catalog No. NC1668447