Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-indazole-3-carboxylic acid, Thermo Scientific™

Catalog No. p-4509663
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 4498-67-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
InChI Key BHXVYTQDWMQVBI-UHFFFAOYSA-N
Synonym indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
PubChem CID 78250
IUPAC Name 1H-indazole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=NN2)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1H-Indazole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.