Learn More
Thomas Scientific 1mL Aspirating Pipet, Bulk Packed, Sterile
SDP
Supplier:  Thomas Scientific 1222B06
Encompass
CELLTREAT offers disposable Aspirating Pipets in 1ml, 2ml and 5ml sizes. Aspirating pipets are supplied without graduations or plugs and are available individually wrapped or bulk-packed. CELLTREAT Scientific Products include a comprehensive "Quality Certificate" with every case of product. The CELLTREAT "Quality Certificate" includes the part number, lot number, sterility assurance level and dosage, endotoxin/pyrogen-free assurance, non-cytotoxicity assurance and RNase-/DNase-free assurance.
Catalog No. 50-146-1454
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.