Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B5322200MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

159622-10-3

Molecular Formula

C11H17NO4

Molecular Weight (g/mol)

227.26

MDL Number

MFCD11042412

InChI Key

RFAQWADNTLIWMG-UHFFFAOYNA-N

Synonym

(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-ethenylcyclopropanecarboxylic Acid

PubChem CID

10376443

IUPAC Name

1-{[(tert-butoxy)carbonyl]amino}-2-ethenylcyclopropane-1-carboxylic acid

SMILES

CC(C)(C)OC(=O)NC1(CC1C=C)C(O)=O

Specifications

Quantity

200 mg

Formula Weight

227.26

Percent Purity

≥98.0% (GC)

Physical Form

Crystalline Powder

Chemical Name or Material

(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic Acid

TSCA

No

Recommended Storage

Refrigerator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.