Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7124040 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 159622-10-3
Molecular Formula C11H17NO4
Molecular Weight (g/mol) 227.26
MDL Number MFCD11042412
InChI Key RFAQWADNTLIWMG-UHFFFAOYNA-N
Synonym (1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-ethenylcyclopropanecarboxylic Acid
PubChem CID 10376443
IUPAC Name 1-{[(tert-butoxy)carbonyl]amino}-2-ethenylcyclopropane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CC1C=C)C(O)=O

Specifications

Quantity 200 mg
Formula Weight 227.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic Acid
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.