Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%, Thermo Scientific Chemicals

Catalog No. p-4602061
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle

CAS: 1517-82-4 | C17H26O2S | 294.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1517-82-4
Molecular Formula C17H26O2S
Molecular Weight (g/mol) 294.46
InChI Key NQICGNSARVCSGJ-DSRNLFJRSA-N
Synonym 1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 101348873
IUPAC Name [(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
SMILES CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C

Specifications

Melting Point 102.0°C to 104.0°C
Color Brown to Yellow or Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation −190° to −204° (20°C, 589nm) (c=2, acetone)
Formula Weight 294.46
Percent Purity 98%
Physical Form Solution
Chemical Name or Material (1R, 2S, 5R)-(-)-Menthyl (S)-p-toluenesulfinate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.