Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-(+)-N,N′-Dimethylcyclohexane-1,2-diamine 98.0+%, TCI America™
SDP

Catalog No. p-7127310 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 87583-89-9
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00671528
InChI Key JRHPOFJADXHYBR-YUMQZZPRSA-N
Synonym 1s,2s-n1,n2-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-1,2-cyclohexanediamine, 1s,2s-+-n,n'-dimethylcyclohexane-1,2-diamine, trans-1s,2s-n,n'-dimethylcyclohexane-1,2-diamine, 1s,2s-+-1,2-bis methylamino cyclohexane, 1s,2s-n,n'-bismethyl-1,2-cyclohexane-diamine, 1s,2s-1-n,2-n-dimethylcyclohexane-1,2-diamine, 1s,2s-n,n'-dimethyl-cyclohexane-1,2-diamine, n,n'-dimethyl-1s,2s-1,2-cyclohexanediamine, trans-1s,2s-n,n-dimethyl-cyclohexane-1,2-diamine
PubChem CID 13822957
IUPAC Name (1S,2S)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CNC1CCCCC1NC

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 100 mg
UN Number 3259
Formula Weight 142.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (1S,2S)-(+)-N,N′-Dimethylcyclohexane-1,2-diamine
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.