Learn More
Perkin Elmer US LLC 1-Slot Demountable Quartz Torch for Avio 200/500
Supplier: Perkin Elmer US LLC N0790131
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions. Included in: Avio 200 Torch Body Assembly (N0790138), Avio 500 Torch Body Assembly (N0810128), Quick Change Torch Module Cyclonic Spray Chamber/Concentric Nebulizer for Avio 200 (N0790607), Quick Change Torch Module Cyclonic Spray Chamber/Concentric Nebulizer for Avio 500 (N0810607), Quick Change Torch Module Scott Spray Chamber/Cross-Flow Nebulizer for Avio 200 (N0790606), Quick Change Torch Module Scott Spray Chamber/Cross-Flow Nebulizer for Avio 500 (N0810606), Cyclonic/Concentric Kit for Avio 200/500 (N0790608) and the Scott/Cross-Flow II Kit (N0790609) for Avio 200/500.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.