Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzothiadiazol-5-amine, 97%, Thermo Scientific™

Catalog No. CC09214EA
Click to view available options
:
Amber Glass Bottle; 10g

Specifications

CAS 874-37-3
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H5N3S
MDL Number MFCD00185826
Quantity 10g
Synonym benzo c 1,2,5 thiadiazol-5-amine, 5-amino-2,1,3-benzothiadiazole, 5-aminobenzo-2,1,3-thiadiazole, benzo c 1,2,5-thiadiazole-5-ylamine, 5-aminobenzo 2,1,3 thiadiazole, 2,1,3-benzothiadiazol-5-ylamine, benzo 1,2,5 thiadiazol-5-ylamine
InChI Key JRJPKRSKPOCUEV-UHFFFAOYSA-N
SMILES NC1=CC2=NSN=C2C=C1
IUPAC Name 2,1,3-benzothiadiazol-5-amine
Molecular Weight (g/mol) 151.19
PubChem CID 314313
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazol-5-amine
Show More Show Less
Hazard Category Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.