Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzoxadiazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC09704DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 32863-33-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H4N2O2
Quantity 1g
MDL Number MFCD01200350
Synonym 2,1,3-benzoxadiazole-5-carboxaldehyde, benzo c 1,2,5 oxadiazole-5-carbaldehyde, 5-formylbenzofurazan, pubchem14542, benzofurazan-5-carboxaldehyde, 5-formyl-2,1,3-benzoxadiazole, benzo 2,1,3 oxadiazol-5-carbaldehyde, benzo 1,2,5 oxadiazole-5-carbaldehyde, benzo 2,1,3 oxadiazole-5-carbaldehyde, 5-benzofurazancarboxaldehyde 8ci ; 5-formyl-2,1,3-benzoxadiazole; 5-formylbenzofurazan; benzo 2,1,3 oxadiazole-5-carboxaldehyde
InChI Key STVDCFOBQWMSHN-UHFFFAOYSA-N
SMILES O=CC1=CC2=NON=C2C=C1
IUPAC Name 2,1,3-benzoxadiazole-5-carbaldehyde
Molecular Weight (g/mol) 148.12
PubChem CID 2776296
Formula Weight 148.12
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole-5-carbaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.