Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Adamantyl)propan-2-ol, 97%, Thermo Scientific™

Catalog No. NRB01891DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 775-64-4
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.32
MDL Number MFCD00167855
InChI Key WBKAUEBLTWRERU-UHFFFAOYSA-N
Synonym 2-adamantan-1-yl-propan-2-ol, 2-1-adamantyl propan-2-ol, 2-adamantan-1-yl propan-2-ol, 2-1-adamantyl-2-propanol, 2-adamantanylpropan-2-ol, pubchem14445, maybridge1_005546, 2-1-adamantly-2-propanol, 2-1-adamantyl-propan-2-ol
PubChem CID 300814
SMILES CC(C)(O)C12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 2-(1-adamantyl)propan-2-ol
Formula Weight 194.32
Percent Purity 97%
Chemical Name or Material 2-(1-Adamantyl)propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.