Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Cyclohexenyl)ethylamine 98.0+%, TCI America™
SDP

Catalog No. p-7124824 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 3399-73-3
Molecular Formula C8H15N
Molecular Weight (g/mol) 125.215
MDL Number MFCD00012058
InChI Key IUDMXOOVKMKODN-UHFFFAOYSA-N
Synonym 2-1-cyclohexenyl ethylamine, 1-cyclohexene-1-ethanamine, 2-cyclohex-1-en-1-yl ethanamine, cyclohexenylethylamine, 1-cyclohexen-1-ylethylamine, 2-1-cyclohexen-1-yl ethylamine, cyclohex-1-ene-1-ethylamine, 2-cyclohex-1-en-1-yl ethan-1-amine, 2-cyclohexen-1-yl ethanamine, .beta.-1-cyclohexenyl ethylamine
PubChem CID 76938
IUPAC Name 2-(cyclohexen-1-yl)ethanamine
SMILES C1CCC(=CC1)CCN

Specifications

Color Yellow
Boiling Point 84°C
Quantity 25 mL
UN Number 2733
Formula Weight 125.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(1-Cyclohexenyl)ethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.