Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. p-7119509 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 371-27-7
Molecular Formula C6H9F3O2
Molecular Weight (g/mol) 170.131
MDL Number MFCD00077609
InChI Key DEXWRCYOMLUJRF-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl butyrate, 2,2,2-trifluoroethylbutyrate, butanoic acid, 2,2,2-trifluoroethyl ester, 2-tfb, butyric acid, tfe, trifluoroethyl butyrate, trifluoroethyl butanoate, acmc-209ir0, butanoic acid,2,2,2-trifluoroethyl ester
PubChem CID 193290
IUPAC Name 2,2,2-trifluoroethyl butanoate
SMILES CCCC(=O)OCC(F)(F)F

Specifications

Color Colorless
Boiling Point 113°C
Quantity 5 g
UN Number 3272
Formula Weight 170.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Butyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.