Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Dichlorophenoxy)propionic acid, 98%, Thermo Scientific™

Catalog No. p-7027526
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 120-36-5
Molecular Formula C9H8Cl2O3
Molecular Weight (g/mol) 235.06
MDL Number MFCD00002645
InChI Key MZHCENGPTKEIGP-UHFFFAOYNA-N
Synonym dichlorprop, 2-2,4-dichlorophenoxy propanoic acid, 2-2,4-dichlorophenoxy propionic acid, hormatox, polymone, dichloroprop, polytox, kildip, celatox-dp, herbizid dp
PubChem CID 8427
ChEBI CHEBI:75370
IUPAC Name 2-(2,4-dichlorophenoxy)propanoic acid
SMILES CC(OC1=CC=C(Cl)C=C1Cl)C(O)=O

Specifications

Melting Point 112°C to 116°C
Odor Odorless
Quantity 100 g
Beilstein 2213812
Merck Index 14,3078
Formula Weight 235.07
Percent Purity 98%
Chemical Name or Material 2-(2,4-Dichlorophenoxy)propionic acid
Hazard Category H302+H312-H315-H318-H351
Hazard Statement GHS H Statement
H318-H302-H312-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-390-5
RTECSNumber UF1050000
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.