Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, MilliporeSigma™

Catalog No. p-3176653 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Density 0.69g/cm3 (20°C)
Quantity 1 L
Explosion Limit 1 to 5.5%(V)
Ignition Point 410°C
Packaging Glass Bottle
Linear Formula CH3C(CH3)2CH2CH(CH3)CH3
Vapor Pressure 51 hPa (20°C)
Formula Weight 114.23
Chemical Name or Material 2, 2, 4-Trimethylpentane, SeccoSolv™, Dried
Recommended Storage Store at +15°C to +25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.