Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2′-Bis(hydroxymethyl)diphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. p-7119178 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 10038-40-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD00191613
InChI Key VRVKKKKXKVCPEW-UHFFFAOYSA-N
Synonym 2,2′C-Oxydi(benzyl Alcohol)
PubChem CID 611393
IUPAC Name [2-[2-(hydroxymethyl)phenoxy]phenyl]methanol
SMILES C1=CC=C(C(=C1)CO)OC2=CC=CC=C2CO

Specifications

Melting Point 99°C
Color White
Quantity 1 g
Formula Weight 230.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bis(hydroxymethyl)diphenyl Ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.