Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoroethylamine 98.0+%, TCI America™
SDP

Catalog No. p-7128443 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 430-67-1
Molecular Formula C2H5F2N
Molecular Weight (g/mol) 81.066
MDL Number MFCD00798137
InChI Key OVRWUZYZECPJOB-UHFFFAOYSA-N
PubChem CID 136274
IUPAC Name 2,2-difluoroethanamine
SMILES C(C(F)F)N

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
UN Number 2733
Formula Weight 81.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoroethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.