Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-(Methylamino)ethyl)pyridine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. p-159319
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5579-84-0
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.114
MDL Number MFCD00012813
InChI Key XVDFMHARQUBJRE-UHFFFAOYSA-N
Synonym betahistine dihydrochloride, betahistine 2hcl, betahistine hydrochloride, serc, antivom, betaserc, microser, betahistine hcl, 2-pyridineethanamine, n-methyl-, dihydrochloride, unii-49k58smz7u
PubChem CID 68643
IUPAC Name N-methyl-2-pyridin-2-ylethanamine;dihydrochloride
SMILES CNCCC1=CC=CC=N1.Cl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 150°C to 154°C
Color White to Yellow
pH 2 to 4 (2% soln.)
Loss on Drying 1% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C5H4N)CH2CH2N(H)CH3·2HCl
Quantity 1 g
Beilstein 22, 434
Merck Index 15, 1180
Formula Weight 209.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-(Methylamino)ethyl)pyridine dihydrochloride, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-966-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.