Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3,4,5,6-Pentabromotoluene, ≥98.0% (GC); 98.0-102.0% (argentometric after oxygen combustion), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
50 mg
Description
Featured Industries: Environmental
Specifications
Specifications
Quantity | 50 mg |
CAS | 87-83-2 |
Type | Analytical Standard |
Linear Formula | C6 Br5 CH3 |
MDL Number | MFCD00000060 |
Synonym | Pentabromotoluene |
Formula Weight | 486.62 |
Percent Purity | ≥98.0% (GC); 98.0 to 102.0% (argentometric after oxygen combustion) |
Grade | Analytical Standard |
Melting Point | 285°C to 286°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction