Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3,5-Triphenyltetrazolium Chloride 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
25 g
Specifications
Specifications
Color | White-Yellow |
Quantity | 1 g |
Formula Weight | 334.81 |
Percent Purity | ≥98.0% (T) |
Physical Form | Powder |
Chemical Name or Material | 2,3,5-Triphenyltetrazolium Chloride |
Safety and Handling
EINECSNumber | (5)-3612 |
RTECSNumber | XF8100000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction