Learn More
Invivogen 2'3'-CGAMP
SDP
Supplier:  Invivogen TLRLNACGA23
Encompass
2’3’-cGAMP (cyclic [G(2’,5’)pA(3’,5’)p]) is produced in mammalian cells by cGAS (cGAMP synthase) in response to double-stranded DNA in the cytoplasm. 2’3’-cGAMP is also referred to as “noncanonical“ cGAMP due to the presence of the atypical 2’-5’ phosphodiester linkage between the guanosine and the adenosine. Similar to the canonical 3’3’-cGAMP, 2’3’-cGAMP serves as a second messenger to activate innate immune responses by binding to STING (stimulator of IFN genes) and subsequently inducing the TBK1-IRF3-dependent production of IFN-β [1]. Structural and functional studies revealed that noncanonical 2’3’-cGAMP is distinct from the canonical 3’3’-cGAMP produced by bacteria [2,3]. 2'3'-cGAMP binds to STING with a much greater affinity than 3'3'-cGAMP [1]. Certain variants of STING are able to distinguish between noncanonical and canonical cGAMP [2]
Catalog No. NC1094221
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.