Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3′-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine 97.0+%, TCI America™
SDP

Catalog No. p-7125866 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 1443049-84-0
Molecular Formula C27H18ClN3
Molecular Weight (g/mol) 419.912
InChI Key PGIGQFHBEHJSQN-UHFFFAOYSA-N
PubChem CID 91972125
IUPAC Name 2-[4-(3-chlorophenyl)phenyl]-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=C(C=C3)C4=CC(=CC=C4)Cl)C5=CC=CC=C5

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 200 mg
Formula Weight 419.91
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3′-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.