Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-(3-Chlorophenyl)-1,3-thiazol-4-yl)methanamine hydrochloride monohydrate, 97%, Thermo Scientific™

Catalog No. MO01298DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 690632-12-3
Molecular Formula C10H10Cl2N2S
Molecular Weight (g/mol) 261.164
MDL Number MFCD06738410
InChI Key DBEFCUHDXFEPSJ-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl thiazol-4-yl methylamine hydrochloride, c-2-3-chloro-phenyl-thiazol-4-yl-methylamine hydrochloride, 2-3-chlorophenyl-1,3-thiazol-4-yl methane amine hydrochloride 1-hydrate, 4-aminomethyl-2-3-chlorophenyl thiazole hcl, 2-3-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-3-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-3-chloro-phenyl-thiazol-4-ylmethyllamine hcl, 2-3-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 1-2-3-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
PubChem CID 24229776
IUPAC Name [2-(3-chlorophenyl)-1,3-thiazol-4-yl]methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)C2=NC(=CS2)CN.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 279.2
Percent Purity 97%
Chemical Name or Material [2-(3-Chlorophenyl)-1, 3-thiazol-4-yl]methanamine hydrochloride monohydrate
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.