Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromo-3-phenylpropiophenone, 98%, Thermo Scientific™

Catalog No. p-4812664
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 611-91-6
Molecular Formula C15H12Br2O
Molecular Weight (g/mol) 368.07
MDL Number MFCD00017861
InChI Key LYAGBKGGYRLVTR-UHFFFAOYNA-N
PubChem CID 95342
IUPAC Name 2,3-dibromo-1,3-diphenylpropan-1-one
SMILES BrC(C(Br)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 156°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CH(Br)C(O)C6H5
Quantity 1 g
Synonym chalcone dibromide, 2,3-dibromo-1,3-diphenyl-1-propanone, benzalacetophenone dibromide, 2,3-dibromo-3-phenylpropiophenone, 1-propanone, 2,3-dibromo-1,3-diphenyl, propiophenone, 2,3-dibromo-3-phenyl, 1-propanone,2,3-dibromo-1,3-diphenyl, dibromodiphenylpropanone, benzal acetophenone dibromide, propiophenone,3-dibromo-3-phenyl
Solubility Information Solubility in water: negligible
Formula Weight 368.07
Percent Purity 98%
Chemical Name or Material 2, 3-Dibromo-3-phenylpropiophenone
Hazard Category GHS Signal Word: Danger
EINECSNumber 210-282-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.