Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7127662 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50-45-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
InChI Key QAOJBHRZQQDFHA-UHFFFAOYSA-N
Synonym benzoic acid, 2,3-dichloro, 2,3-dichloro benzoic acid, unii-cgp39qv5wf, benzoic acid, dichloro, 2,3-dichorobenzoic acid, cgp39qv5wf, dichlorobenzoic acid, lamotrigine impurity e, pubchem8198, acmc-209kmq
PubChem CID 5765
IUPAC Name 2,3-dichlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C(=O)O

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorobenzoic Acid
EINECSNumber (3)-0062&(3)-1410&(3)-1424
RTECSNumber DG6475000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.