Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 2',3'-Dideoxyinosine, 98%

Catalog No. p-7042982
Click to view available options
Quantity:
250 mg
500 mg
1 g

2?,3?-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,3′-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

Solubility
Soluble in DMSO or methanol

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 69655-05-6
Molecular Formula C10H12N4O3
Molecular Weight (g/mol) 236.23
MDL Number MFCD00077728
InChI Key BXZVVICBKDXVGW-JHPDDGAFNA-N
Synonym didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi
PubChem CID 50599
ChEBI CHEBI:490877
IUPAC Name 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O

Specifications

Color White
Quantity 250 mg
Beilstein 3619529
Merck Index 143098
Solubility Information Soluble in DMSO or methanol
Formula Weight 236.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2',3'-Dideoxyinosine
RTECSNumber NM7460700
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.