Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,4,6-Trimethylbenzene-1-thiol, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.0240g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)3C6H2SH |
Quantity | 10 g |
Formula Weight | 152.26 |
Percent Purity | 97% |
Chemical Name or Material | 2, 4, 6-Trimethylbenzene-1-thiol |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement: Harmful if inhaled. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | GHS P Statement: IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction