Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Biphenylyl)amino-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. p-7123528 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 897671-69-1
Molecular Formula C27H23N
Molecular Weight (g/mol) 361.49
MDL Number MFCD16036286
InChI Key QRMLAMCEPKEKHS-UHFFFAOYSA-N
PubChem CID 20730584
IUPAC Name N-{[1,1'-biphenyl]-4-yl}-9,9-dimethyl-9H-fluoren-2-amine
SMILES CC1(C)C2=CC=CC=C2C2=C1C=C(NC1=CC=C(C=C1)C1=CC=CC=C1)C=C2

Specifications

Melting Point 147°C
Color White
Quantity 1 g
Synonym N-([1,1′C-biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine
Formula Weight 361.49
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Biphenylyl)amino-9,9-dimethylfluorene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.