Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chloro-3-nitrobenzoyl)benzoic Acid 97.0+%, TCI America™
SDP

Catalog No. p-7124742 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 85-54-1
Molecular Formula C14H8ClNO5
Molecular Weight (g/mol) 305.67
MDL Number MFCD00007082
InChI Key RITAQDHCJBLSSL-UHFFFAOYSA-N
Synonym 2-4-chloro-3-nitrobenzoyl benzoic acid, benzoic acid, 2-4-chloro-3-nitrobenzoyl, 3'-nitro-4'-chlorobenzoylbenzoic acid, benzoic acid, o-4-chloro-3-nitrobenzoyl, 2-4-chloro-3-nitrophenyl carbonyl benzoic acid, acmc-209q6m, ksc496o4t, o-4-chloro-3-nitrobenzoyl benzoic acid, 2-4-chloro-3-nitro-benzoyl benzoic acid, 2-4'-chloro-3'-nitrobenzoyl benzoic acid
PubChem CID 66562
IUPAC Name 2-(4-chloro-3-nitrobenzoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 g
Formula Weight 305.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Chloro-3-nitrobenzoyl)benzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.