Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,4-Dibromophenol 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
Melting Point | 39°C |
Color | White-Yellow |
Boiling Point | 239°C |
Quantity | 25 g |
UN Number | 2811 |
Formula Weight | 251.91 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2,4-Dibromophenol |
Safety and Handling
EINECSNumber | (3)-0907 |
RTECSNumber | SK8010000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction