Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. p-7119220 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 23915-07-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD00011649
InChI Key IBLMYGXJKQIGSN-UHFFFAOYSA-N
Synonym 2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911
PubChem CID 90297
IUPAC Name 1-(bromomethyl)-2,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)F)CBr

Specifications

Color Yellow
Quantity 5 g
UN Number 2920
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzyl Bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.