Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-3-furoyl chloride, 90%, Thermo Scientific™

Catalog No. p-4391325
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 15139-39-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
InChI Key NOEQXZGBSMGWCT-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furoyl chloride, 2,4-dimethylfuran-3-carboxylic chloride, 3-furancarbonyl chloride, 2,4-dimethyl-9ci
PubChem CID 13019985
IUPAC Name 2,4-dimethylfuran-3-carbonyl chloride
SMILES CC1=COC(=C1C(=O)Cl)C

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.58
Percent Purity 90%
Chemical Name or Material 2, 4-Dimethyl-3-furoyl chloride
Hazard Category GHS Signal Word: Danger
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.