Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyloxazole 98.0+%, TCI America™
SDP

Catalog No. p-7127901 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 7208-05-1
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.12
MDL Number MFCD00234124
InChI Key PSOZJOZKEVZLKZ-UHFFFAOYSA-N
PubChem CID 138961
IUPAC Name 2,4-dimethyl-1,3-oxazole
SMILES CC1=COC(=N1)C

Specifications

Color Yellow
Boiling Point 108°C
Quantity 1 g
UN Number 1993
Formula Weight 97.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyloxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.