Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methoxyphenoxy)ethylamine, Thermo Scientific™

Catalog No. RF03346DA
Click to view available options
Quantity:
250mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 50800-92-5
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052978
InChI Key YXRCDWIZAGWUBL-UHFFFAOYSA-N
Synonym 2-4-methoxyphenoxy ethylamine, 2-4-methoxyphenoxy ethanamine, 2-4-methoxyphenoxy-1-ethanamine, 1-2-aminoethoxy-4-methoxybenzene, 2-4-methoxy-phenoxy-ethylamine, 2-4-methoxyphenoxy ethan-1-amine, ethanamine, 2-4-methoxyphenoxy, maybridge3_000601, 2-4-methoxy-phenoxy ethylamine
PubChem CID 1797793
IUPAC Name 2-(4-methoxyphenoxy)ethanamine
SMILES COC1=CC=C(C=C1)OCCN

Specifications

Packaging Amber Glass Bottle
Linear Formula NH2(CH2)2OC6H4OCH3
Quantity 1 g
Formula Weight 167.21
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(4-Methoxyphenoxy)ethylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.