Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Piperidinedione, 97%, Thermo Scientific Chemicals

Catalog No. p-4380107
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50607-30-2
InChI Key RDNZDMDLRIQQAX-UHFFFAOYSA-N
PubChem CID 10887863
IUPAC Name piperidine-2,4-dione
SMILES C1CNC(=O)CC1=O

Specifications

Melting Point 99.1°C to 100.1°C
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C5H7NO2
Quantity 1 g
Synonym 2,4-piperidinedione, 2,4-piperadinedione, 2,4-dioxopiperidine, 2,4-diketopiperidine, piperidin-2,4-dione, 2,4 dioxopiperdine, 2, 4-dioxopiperidine, pubchem22543, acmc-209ko8
Molecular Weight (g/mol) 113.12
Formula Weight 113.12
Percent Purity 97%
Chemical Name or Material 2, 4-Piperidinedione
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.